missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other TBEV gE Protein
A cDNA sequence encoding the TBEV gE was constructed and used to recombinantly synthesize the protein.
€ 238.00 - € 1515.00
Specifications
Name | TBEV gE Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15924419
|
enQuireBio™
QP13698-100UG |
100 μg |
€ 238.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944419
|
enQuireBio™
QP13698-500UG |
500 μg |
€ 910.00
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934419
|
enQuireBio™
QP13698-1MG |
1 mg |
€ 1515.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TBEV gE Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Protein is >90% pure as determined by SDS- PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-MES pH 6.5, 8M urea, 200mM NaCl and 0.05% Tween-20. |