missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa L-ornithine N(5)-monooxygenase Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) L-ornithine N(5)-monooxygenase, was expressed in the hosts and tags indicated. option, ab
€ 309.00 - € 3405.00
Specifications
Accession Number | NP_251076.1 |
---|---|
Gene ID (Entrez) | 882167 |
Name | P. aeruginosa L-ornithine N(5)-monooxygenase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030121
|
enQuireBio™
QP7676-EC-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050121
|
enQuireBio™
QP7676-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040121
|
enQuireBio™
QP7676-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060121
|
enQuireBio™
QP7676-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070121
|
enQuireBio™
QP7676-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080121
|
enQuireBio™
QP7676-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_251076.1 | |
P. aeruginosa L-ornithine N(5)-monooxygenase Protein | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
882167 | |
Research Use Only | |
pvdA | |
Recombinant Protein | |
E. coli | |
MTQATATAVV HDLIGVGFGP SNIALAIALQ ERAQAQGALE VLFLDKQGDY RWHGNTLVSQ SELQISFLKD LVSLRNPTSP YSFVNYLHKH DRLVDFINLG TFYPCRMEFN DYLRWVASHF QEQSRYGEEV LRIEPMLSAG QVEALRVISR NADGEELVRT TRALVVSPGG TPRIPQVFRA LKGDGRVFHH SQYLEHMAKQ PCSSGKPMKI AIIGGGQSAA EAFIDLNDSY PSVQADMILR ASALKPADDS PFVNEVFAPK FTDLIYSREH AERERLLREY HNTNYSVVDT DLIERIYGVF YRQKVSGIPR HAFRCMTTVE RATATAQGIE LALRDAGSGE LSVETYDAVI LATGYERQLH RQLLEPLAEY LGDHEIGRDY RLQTDERCKV AIYAQGFSQA SHGLSDTLLS VLPVRAEEIS GSLYQHLKPG TAARALHEHA LAS | |
Greater than 90% as determined by SDS-PAGE. |