Learn More
enQuireBio™ Recombinant Parvovirus Parvovirus VP2 Protein
Recombinant Protein
Brand:  enQuireBio™ QP10489-2mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15944018
  • € 3800.00 / 2mg
Estimated Shipment: 25-06-2024
to see stock.
Specifications
Parvovirus Parvovirus VP2 Protein
Research Use Only
Recombinant Protein
Human
Tris NaCl pH7.9.
2 mg
Untested
Parvovirus
Native
>90% as determined by SDS-PAGE.