missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig ETFDH Protein

Product Code. 15919279
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15919279

Brand: enQuireBio™ QP6003ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Sus scrofa (Pig) ETFDH, was expressed in the hosts and tags indicated.

Specifications

Name ETFDH Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ETFDH
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Pig
Species E. coli
Protein Tag His-SUMO
Sequence SSTCKVPRIT THYTIYPRDQ DKRWEGVNME RFAEEADVVI VGAGPAGLSA ATRLKQLAAQ HEKDLRVCLV EKAAHIGAHT LSGACLDPRA FEELFPDWKE KGAPLNTPVT EDRFGILTEK YRIPVPILPG LPMNNHGNYV VRLGHLVSWM GEQAEALGVE VYPGYAAAEI LFHEDGSVKG IATNDVGIQK DGAPKTTFER GLELHAKVTI FAEGCHGHLA KQLYKKFDLR ANCEPQTYGI GLKELWVIDE KKWKPGRVDH TVGWPLDRHT YGGSFLYHLN EGEPLLALGF VVGLDYQNPY LSPFREFQRW KHHPSIKPTL EGGKRIAYGA RALNEGGFQS IPKLTFPGGL LIGCSPGFMN VPKIKGTHTA MKSGTLAAES IFNQLTSENL QSKTIGLHVT EYEDNLKNSW VWKELYSVRN IRPSCHGILG VYGGMIYTGI FYWIFRGMEP WTLKHKGSDS DQLKPAKDCT PIEYPKPDGQ ISFDLLSSVA LSGTNHEHDQ PAHLTLKDDS VPVNRNLSIY DGPEQRFCPA GVYEFVPLEQ GDGFRLQINA QNCVHCKTCD IKDPSQNINW VVPEGGGGPA YNGM
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 85.3% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.