Learn More
enQuireBio™ Recombinant Porcine Thrombin Protein
Product Code. 15915448
Click to view available options
Quantity:
1000 IU
2000 IU
500 U
Unit Size:
1000 units
2000 units
500 units
Product Code. 15915448
Brand: enQuireBio™ QP108781000U
A cDNA sequence encoding the Thrombin was constructed and used to recombinantly synthesize the protein.
Specifications
Name Thrombin Protein
Quantity 1000 IU
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity One unit is defined as the amount of enzyme needed to cleave 1 mg of fusion protein in 16 hours to 95% completion at 20C in a buffer containing 25mM Tris-HCl, pH 8.4, 150mM NaCl, and 2.5mM CaCl2.
Product Type Recombinant Protein
Cross Reactivity Porcine
Species Porcine
Protein Tag Untagged
Buffer Lyophilized Powder from 20mM PBS, pH 7.4 containing 0.9% NaCl.