Learn More
R&D Systems™ Recombinant Porcine Tie-2 Fc Chimera Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Porcine Tie-2 Fc Chimera (Catalog # 10488-T2) is immobilized at 0.5 μg/mL (100 μL/well), Recombinant Human Angiopoietin-2 His-tag (Catalog # 623-AN ) binds with an ED50 of 0.3-2.7 ng/mL.
Specifications
Specifications
| Accession Number | XP_020918337.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 7010 |
| Molecular Weight (g/mol) | M.W.-observed: 115-133 kDa, under reducing conditions, M.W.-theoretical: 107 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived porcine Tie-2 protein Porcine Tie-2 (Ala23-Lys746) (N-terminus) Accession # XP_020918337.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK, TEK tyrosine kinase, endothelial, Tie2, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 |
| Show More |