Learn More
R&D Systems™ Recombinant Rabbit IL-21 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
missing translation for 'unitSize'
25µg
Description
The Recombinant Rabbit IL-21 Protein is derived from E. coli. The Recombinant Rabbit IL-21 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | XP_008266193 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 100344172 |
| Molecular Weight (g/mol) | 15.5 kDa |
| Name | IL-21 |
| Quantity | 25 μg |
| Source | E. coli-derived rabbit IL-21 protein His18-Arg150,with an N-terminal Met, |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Monomer |
| Conjugate | Unconjugated |
| Show More |