missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit IL2 / Interleukin-2 Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6218-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001156652.1 | |
IL2 / Interleukin-2 Protein | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
APTSSSTKET QEQLDQLLLD LQVLLKGVND YKNSKLSRML TFKFYMPKKV TELKHLQCLE EELKPLEEVL NLAQGKNSHG GNTRESISNI NVTVLKLKGS ETFMCEYDET VTIVEFLNRW ITFCQSIISA SSS | |
Greater than 90% as determined by SDS-PAGE. |
100302458 | |
500 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |