missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rainbow Trout GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10630-10ug
Additional Details : Weight : 0.01000kg
Specifications
GH Protein | |
Research Use Only | |
Somatotropin Rainbow Trout (Oncorhynchus mykiss) Recombinant is biologically active in PDF-P1 3B9 cells stable transfected with rabbit GH receptors, though its activity is about 10 fold lower than that of human GH. | |
Rainbow Trout | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.5% NaHCO3. Adjusted to pH 8. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
AIENQRLFNIAVSRVQHLHLLAQKMFNDFDGTLLPDERRQLNKIFLLDFCNSDSIVSPVDKHETQKSSVLKLLHISFRLIESWEYPSQTLIISNSLMVRNANQISEKLSDLKVGINLLITGSQDGVLSLDDNDSQQLPPYGNYYQNLGGDGNVRRNYELLACFKKDMHKVETYLTVAKCRKSLEANCTL | |
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |