missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Caprin-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Caprin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7683-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001012185.2 | |
Caprin-1 Protein | |
Research Use Only | |
CAPRIN1 | |
Recombinant Protein | |
E. coli | |
PSATSHSGSG SKSSGPPPPS GSSGSEAAAG AAAPASQHPA TGTGAVQTEA MKQILGVIDK KLRNLEKKKG KLDDYQERMN KGERLNQDQL DAVSKYQEVT NNLEFAKELQ RSFMALSQDI QKTIKKTARR EQLMREEAEQ KRLKTVLELQ YVLDKLGDDD VRTDLKQGLS GVPILSEEEL SLLDEFYKLV DPERDMSLRL NEQYEHASIH LWDLLEGKEK PVCGTTYKAL KEIVERVFQS NYFDSTHNHQ NGLCEEEEAA SAPIVEDQVA EAEPEPTEEY TEQSEVESTE YVNRQFMAET QFSSGEKEQV DEWAVETVEV VNSLQQQPQA ASPSVPEPHS LTPVAQSDPL VRRQRVQDLM AQMQGPYNFI QDSMLDFENQ TLDPAIVSAQ PMNPTQNMDM PQLVCPQVHS ESRLAQSNQV PVQPEATQVP LVSSTSEGYT ASQPLYQPSH AAEQRPQKEP IDQIQATISL NTEQTTASSS LPAASQPQVF QAGASKPLHS SGINVNAAPF QSMQTVFNMN APVPPVNEPE TLKQQSQYQA SYNQSFSSQP HQVEQTELQQ DQLQTVVGTY HGSQDQPHQV PGNHQQPPQQ STGFPRSSQP YYNSRGVSRG GSRGARGLMN GYRGPANGFR GGYDGYRSSF SNTPNSGYTQ SQFNAPRDYS GYQRDGYQQN FKRGSGQSGP RGAPRGRGGP PRPNRGMPQM NTQQVN | |
Greater than 90% as determined by SDS-PAGE. |
362173 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |