missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Carboxypeptidase B Protein

Product Code. 15964198
Click to view available options
Quantity:
5 mg
15 mg
50 mg
Unit Size:
15mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 15964198

Brand: enQuireBio™ QP1054415mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Carboxypeptidase B was constructed and used to recombinantly synthesize the protein.

Specifications

Name Carboxypeptidase B Protein
Quantity 15 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity 170 units/mg protein.
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag Untagged
Sequence MLLLLALVSV ALAHASEEHF DGNRVYRVSV HGEDHVNLIQ ELANTKEIDF WKPDSATQVK PLTTVDFHVK AEDVADVENF LEENEVHYEV LISNVRNALE SQFDSHTRAS GHSYTKYNKW ETIEAWIQQV ATDNPDLVTQ SVIGTTFEGR NMYVLKIGKT RPNKPAIFID CGFHAREWIS PAFCQWFVRE AVRTYNQEIH MKQLLDELDF YVLPVVNIDG YVYTWTKDRM WRKTRSTMAG SSCLGVRPNR NFNAGWCEVG ASRSPCSETY CGPAPESEKE TKALADFIRN NLSTIKAYLT IHSYSQMMLY PYSYDYKLPE NYEELNALVK GAAKELATLH GTKYTYGPGA TTIYPAAGGS DDWSYDQGIK YSFTFELRDT GFFGFLLPES QIRQTCEETM LAVKYIANYV REHLY
Buffer The protein was lyophilized with 100mM NaCl and 20mM Tris pH 7.5.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.