missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CDNF Protein

A cDNA sequence encoding the CDNF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10549-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934218

  • € 120.00 / 5µg

Estimated Shipment: 06-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

CDNF Protein
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
QGLEAGVRSR ADCEVCKEFL NRFYNSLLTR GIDFSVDTIE EELISFCADT KGKENRLCYY LGATKDSATK ILGEVTRPMS VHMPTVKICE KLKKMDSQIC ELKYEKKLDL ESVDLWKMRV AELKQILHSW GEECRACAEK HDYVNLIKEL APKYVETRPQ TEL
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
CDNF Rat is able to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons when immobilized at 5-25 microg/mL on a nitrocellulose-coated microplate.
Rat
Untagged
CDNF protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers