missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Complement C3 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Complement C3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9660-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_058690.2 | |
Complement C3 Protein | |
Research Use Only | |
C3 | |
Recombinant Protein | |
Yeast | |
SVQLMERRMD KAGQYTDKGL RKCCEDGMRD IPMPYSCQRR ARLITQGESC LKAFMDCCNY ITKLREQHRR DHVLGL | |
Greater than 90% as determined by SDS-PAGE. |
24232 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |