missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Complement C3 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Complement C3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9660-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088561

  • € 770.00 / 100µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_058690.2
Complement C3 Protein
Research Use Only
C3
Recombinant Protein
Yeast
SVQLMERRMD KAGQYTDKGL RKCCEDGMRD IPMPYSCQRR ARLITQGESC LKAFMDCCNY ITKLREQHRR DHVLGL
Greater than 90% as determined by SDS-PAGE.
24232
100 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers