missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat CXCL12/SDF-1 alpha Protein

Product Code. 18648625 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18648625

Brand: Novus Biologicals™ NBP2352901mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

SDF1 alpha Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 50-100 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl.
Gene ID (Entrez) 6387
Molecular Weight (g/mol) MolecularWeight-theroretical: 7.9 kDa
Quantity 1 mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of CXCL12/SDF-1 alpha as determined by LAL method.
Gene Alias CXCL12 alpha, CXCL12 chemokine (C-X-C motif) ligand 12, IRH, PBSF, SCYB12, SDF1, SDF-1 alpha, SDF-1 alpha/CXCL12 alpha, SDF1A, SDF1B, TLSF, TPAR1
Gene Symbol CXCL12
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Biologically Active Proteins
Purity or Quality Grade >97 % pure by SDS-PAGE and HPLC
Protein CXCL12/SDF-1 alpha
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.