missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11964-200ug
Weitere Details : Weight : 0.01000kg
Spezifikation
GH Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAADSQTPWL LTFSLLCLLW PQEAGAFPAM PLSSLFANAV LRAQHLHQLA ADTYKEFERA YIPEGQRYSI QNAQAAFCFS ETIPAPTGKE EAQQRTDMEL LRFSLLLIQS WLGPVQFLSR IFTNSLMFGT SDRVYEKLKD LEEGIQALMQ ELEDGSPRIG QILKQTYDKF DANMRSDDAL LKNYGLLSCF KKDLHKAETY LRVMKCRRFA ESSCAF | |
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
200 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
Untagged | |
The protein (1 mg/ml) was lyophilized after extensive dialyses against 5mM phosphate buffer, 5 mg mannitol and 1 mg glycine. |