missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Gremlin 1 / GREM1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Gremlin 1 / GREM1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6118-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030940

Quantity Price
1 € 1715.00 / 500µg
3 € 1600.44 / 500µg
Estimated Shipment
23-10-2020
Add to basket


Specifications

Specifications

NP_062155.1
Gremlin 1 / GREM1 Protein
500 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
50566
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Grem1
Recombinant Protein
E. coli
KKKGSQGAIP PPDKAQHNDS EQTQSPPQPG SRTRGRGQGR GTAMPGEEVL ESSQEALHVT ERKYLKRDWC KTQPLKQTIH EEGCNSRTII NRFCYGQCNS FYIPRHIRKE EGSFQSCSFC KPKKFTTMMV TLNCPELQPP TKKKRVTRVK QCRCISIDLD
Videos
Documents