missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IFNG / Interferon Gamma Protein
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
Specifications
Specifications
| Accession Number | NP_620235.1 |
| Gene ID (Entrez) | 25712 |
| Name | IFNG / Interferon Gamma Protein |
| Quantity | 250 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | IFNG |
| Product Type | Recombinant Protein |
| Cross Reactivity | Rat |
| Species | E. coli |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction