missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IL-10 (Cys167Tyr) Protein
Click to view available options
:
10ug
Description
The Recombinant Rat IL-10 (Cys167Tyr) Protein is derived from E. coli. The Recombinant Rat IL-10 (Cys167Tyr) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 25325 |
| Molecular Weight (g/mol) | 18.7 kDa (monomer) |
| Name | IL-10 (Cys167Tyr) |
| Quantity | 10μg |
| Source | E. coli-derived rat IL-10 protein Ser19-Asn178 (Cys167Tyr) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction