missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IL-21R Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Rat IL-21 R Fc Chimera Protein is derived from NS0. The Recombinant Rat IL-21 R Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 52.2 kDa (monomer) |
| Name | IL-21R |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived rat IL-21 R protein, (N-terminus) Rat IL-21 R (Met1-Pro236) Accession &Num; Q5EBB1 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction