missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat MIF Protein

A cDNA sequence encoding the MIF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12696-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941259

  • € 212.00 / 25µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MIF Protein
Research Use Only
Recombinant Protein
E. coli
MPMFIVNTNV PRASVPEGFL SELTQQLAQA TGKPAQYIAV HVVPDQLMTF SGTSDPCALC SLHSIGKIGG AQNRNYSKLL CGLLSDRLHI SPDRVYINYY DMNAANVGWN GSTFA
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4 and 5% trehalose.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers