missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Nqo2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Nqo2, was expressed in the hosts and tags indicated.
€ 245.00 - € 2645.00
Specifications
Accession Number | NP_001004214.1 |
---|---|
Gene ID (Entrez) | 291084 |
Name | Nqo2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000301
|
enQuireBio™
QP7703-YE-100UG |
100 μg |
€ 770.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040301
|
enQuireBio™
QP7703-YE-500UG |
500 μg |
€ 2195.00
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010301
|
enQuireBio™
QP7703-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030301
|
enQuireBio™
QP7703-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050301
|
enQuireBio™
QP7703-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020301
|
enQuireBio™
QP7703-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001004214.1 | |
Nqo2 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
291084 | |
Research Use Only | |
Nqo2 | |
Recombinant Protein | |
Yeast | |
MAGKKVLLVY AHQEPKSFNG SMKQVAVEEL SKQGCTVTVS DLYTMNFEPR ATRNDVTGAL SNPEVFKYGI EAYEAYKKKA LTSDILEEQR KVQEADLVIF QFPLYWFSVP AILKGWMDRV LCQGFAFDVP GFYDSGFLKD KLALLSFTTG GTAEMYTKAG VNGDFRYFLW PLQHGTLHFC GFKVLAPQIS FGPEVSSEEQ RKVMLASWVQ RLKSIWKEEP IHCTPSWYFQ G | |
Greater than 90% as determined by SDS-PAGE. |