missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Podocalyxin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Podocalyxin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9939-ye-1mg
Additional Details : Weight : 0.01000kg
Spécification
NP_620203.1 | |
Podocalyxin Protein | |
Research Use Only | |
PODXL | |
Recombinant Protein | |
Yeast | |
QDNGNKTDTS DITSIDQNQD KPATNQPSNA TPKSSVQPPT PTSISTSSPD PKATQSSNSS VTTTSDSTTD RTSSSTSTVP TTSNSGQTVS SGGKSSDKIT TALPTTLGPV NASSQPTDLN TSTKLPSTPT TNSTASPHQP VSHSEGQHTT VQSSSASVSS SDNTTLLWIL TTSKPTGTSE GTQPIAISTP GITTPVSTPL QPTGSPGGTE SVPTTEEFTH STSSWTPVVS QGPSTPSSTW TSGSYKLKCD PAIKPHEELL ILNLTRDSFC KGSPPNERFL ELLCHSAKAS FKPAEDSCAL ELAPILDNMS -QAVAVKRIV IETKLSPKAV FELLKDKWDD LTEAGVIDIH LGKEGPPEVN EDRFS | |
Greater than 90% as determined by SDS-PAGE. |
192181 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |