missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat S100G Protein

A cDNA sequence encoding the S100G was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13406-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933469

  • € 2930.00 / 100µg

Estimated Shipment: 03-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

24249
Greater than 90% as determined by SDS-PAGE.
Research Use Only
S100g
Rat
His
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 100mM Phosphate buffer, pH 7.3.
S100G Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSAKKSPEEM KSIFQKYAAK EGDPNQLSKE ELKLLIQSEF PSLLKASSTL DNLFKELDKN GDGEVSYEEF EVFFKKLSQ
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers