missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Semaphorin 3A / SEMA3A Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Semaphorin 3A / SEMA3A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7692-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_059006.1 | |
Semaphorin 3A / SEMA3A Protein | |
Research Use Only | |
Sema3a | |
Recombinant Protein | |
E. coli | |
YANGKNNVPR LKLSYKEMLE SNNVITFNGL ANSSSYHTFL LDEERSRLYV GAKDHIFSFN LVNIKDFQKI VWPVSYTRRD ECKWAGKDIL KECANFIKVL KAYNQTHLYA CGTGAFHPIC TYIEVGHHPE DNIFKLQDSH FENGRGKSPY DPKLLTASLL IDGELYSGTA ADFMGRDFAI FRTLGHHHPI RTEQHDSRWL NDPRFISAHL IPESDNPEDD KVYFFFRENA IDGEHSGKAT HARIGQICKN DFGGHRSLVN KWTTFLKARL ICSVPGPNGI DTHFDELQDV FLMNSKDPKN PIVYGVFTTS SNIFKGSAVC MYSMSDVRRV FLGPYAHRDG PNYQWVPYQG RVPYPRPGTC PSKTFGGFDS TKDLPDDVIT FARSHPAMYN PVFPINNRPI MIKTDVNYQF TQIVVDRVDA EDGQYDVMFI GTDVGTVLKV VSVPKETWHD LEEVLLEEMT VFREPTTISA MELSTKQQQL YIGSTAGVAQ LPLHRCDIYG KACAECCLAR DPYCAWDGSS CSRYFPTAKR RTRRQDIRNG DPLTHCSDLQ HHDNHHGHSL EERIIYGVEN SSTFLECSPK SQRALVYWQF QRRNEDRKEE IRVGDHIIRT EQGLLLRSLQ KKDSGNYLCH AVEHGFMQTL LKVTLEVIDT EHLEELLHKD DDGDGSKTKE MSSSMTPSQK VWYRDFMQLI NHPNLNTMDE FCEQVWKRDR KQRRQRPGHS QGSSNKWKHM QESKKGRNRR THEFERAPRS V | |
Greater than 90% as determined by SDS-PAGE. |
29751 | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |