missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Tnf alpha Protein

Product Code. 15976279
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
5µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15976279 1 mg 1mg
15906289 5 μg 5µg
15986279 20 μg 20µg
15966279 100 μg 100µg
15996279 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15976279 Supplier enQuireBio™ Supplier No. QP55081mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_036807.1
Gene ID (Entrez) 103694380
Name Tnf alpha Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol Tnf
Biological Activity Cytolysis of mouse L929 cells in the presence of Actinomycin D significantly induced by less than 50 pg/ml corresponding to an activity of at least 2.0 x 10^7 units/mg (or a typical ED50 of less than 10 pg/ml).
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag Untagged
Sequence MLRSSSQNSS DKPVAHVVAN HQAEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLIY SQVLFKGQGC PDYVLLTHTV SRFATSYQEK VSLLSAIKSP CPKDTPEGAE LKPWYEPMYL GGVSQLEKGD LLSAEVNLPK YLDITESGQV YFGVIAL
Buffer 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.