missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat VEGF 164 Protein

Product Code. 18620386 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18620386

Brand: Novus Biologicals™ NBP23531010ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

VEGF 165 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human umbilical vein endothelial cells(HUVEC) is less than 5 ng/mL, corresponding to a specific activity of > 2.0 x 10^5 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16612
For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM Tris, 300 mM NaCl, pH 8.8.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) MolecularWeight-theroretical: 38.7 kDa
Quantity 10 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of VEGF 164 AS determined by LAL method.
Gene Symbol VEGFA
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Biologically Active Proteins
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein VEGF 164
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.