missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque IL 13 Protein
A cDNA sequence encoding the IL 13 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 7090.00
Specifications
Name | IL 13 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Rhesus Macaque |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944668
|
enQuireBio™
QP10691-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924668
|
enQuireBio™
QP10691-10UG |
10 μg |
€ 212.00
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934668
|
enQuireBio™
QP10691-1MG |
1 mg |
€ 7090.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL 13 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2?m filtered concentrated solution in PBS, pH 7.4 containing 5% trehalose. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SPSPVPRSTA LKELIEELVN ITQNQKAPLC NGSMVWSINL TAGVYCAALE SLINVSGCSA IEKTQRMLNG FCPHKVSAGQ FSSLRVRDTK IEVAQFVKDL LVHLKKLFRE GRFN | |
Greater than 97.0% as determined by SDS-PAGE and HPLC analyses. |