missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rhesus Macaque IL-13 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
IL-13 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 5 ng/mL, corresponding to a specific activity of > 2.0 x 10^5 IU/mg
Specifications
Specifications
| Accession Number | Q864V6 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4, 3% trehalose. |
| Gene ID (Entrez) | 3596 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 12.6 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of IL-13 AS determined by LAL method. |
| Gene Alias | ALRHMGC116789, BHR1interleukin-13, IL-13 mgC116788, interleukin 13, MGC116786, NC30, P600 |
| Gene Symbol | IL13 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction