Learn More
enQuireBio™ Recombinant Rotavirus Non-structural glycoprotein 1
A DNA sequence encoding the Rotavirus A (isolate Monkey/South Africa/SA11-H96/1958 G3-P5B[2]-I2-R2-C5-M5-A5-N5-T5-E2-H5) (RV-A) (Simian Agent 11 (isolate SI/South Africa/H96/58)) Non-structural glycoprotein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7439-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
YP_002302223.1 | |
Rotavirus Non-structural glycoprotein 1 | |
Research Use Only | |
NSP4 | |
Recombinant Protein | |
E. coli | |
PTMKIALKTS KCSYKVVKYC IVTIFNTLLK LAGYKEQITT KDEIEKQMDR VVKEMRRQLE MIDKLTTREI EQVELLKRIY DKLTVQTTGE IDMTKEINQK NVRTLEEWES GKNPYEPREV TAAM | |
Greater than 90% as determined by SDS-PAGE. |
7011361 | |
500 μg | |
Not determined. | |
Cytoplasmic Domain | |
Rotavirus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |