missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Clumping factor A Protein

A DNA sequence encoding the Staphylococcus aureus Clumping factor A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7758-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030631

Quantity Price
1 € 1715.00 / 500µg
3 € 1600.44 / 500µg
Estimated Shipment
03-11-2020
Add to basket


Specifications

Specifications

WP_001056174.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
clfA
Recombinant Protein
E. coli
GKDITNQLTN VTVGIDSGDT VYPHQAGYVK LNYGFSVPNS AVKGDTFKIT VPKELNLNGV TSTAKVPPIM AGDQVLANGV IDSDGNVIYT FTDYVDTKEN VTANITMPAY IDPENVTKTG NVTLTTGIGS TTANKTVLVD YEKYGKFYNL SIKGTIDQID KTNNTYRQTI YVNPSGDNVI APVLTGNLKP NTDSNALIDQ QNTSIKVYKV DNAADLSESY FVNPENFEDV TNSVNITFPN PNQYKVEFNT PDDQITTPYI VVVNGHIDPN SKGDLALRST LYGYDSRFVW RSMSWDNEVA FNNGSGSGDG IDKPVVPEQP DEPGEIEPIP E
S. aureus Clumping factor A Protein
500 μg
Not determined.
Partial (see sequence information for more details).
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents