missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Clumping factor A Protein

A DNA sequence encoding the Staphylococcus aureus Clumping factor A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7758-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16000631

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_001056174.1
10 μg
Not determined.
Partial (see sequence information for more details).
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
S. aureus Clumping factor A Protein
Research Use Only
clfA
Recombinant Protein
E. coli
GKDITNQLTN VTVGIDSGDT VYPHQAGYVK LNYGFSVPNS AVKGDTFKIT VPKELNLNGV TSTAKVPPIM AGDQVLANGV IDSDGNVIYT FTDYVDTKEN VTANITMPAY IDPENVTKTG NVTLTTGIGS TTANKTVLVD YEKYGKFYNL SIKGTIDQID KTNNTYRQTI YVNPSGDNVI APVLTGNLKP NTDSNALIDQ QNTSIKVYKV DNAADLSESY FVNPENFEDV TNSVNITFPN PNQYKVEFNT PDDQITTPYI VVVNGHIDPN SKGDLALRST LYGYDSRFVW RSMSWDNEVA FNNGSGSGDG IDKPVVPEQP DEPGEIEPIP E
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers