missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Clumping factor A Protein
A DNA sequence encoding the Staphylococcus aureus Clumping factor A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7758-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
WP_001056174.1 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Clumping factor A Protein | |
Research Use Only | |
clfA | |
Recombinant Protein | |
E. coli | |
GKDITNQLTN VTVGIDSGDT VYPHQAGYVK LNYGFSVPNS AVKGDTFKIT VPKELNLNGV TSTAKVPPIM AGDQVLANGV IDSDGNVIYT FTDYVDTKEN VTANITMPAY IDPENVTKTG NVTLTTGIGS TTANKTVLVD YEKYGKFYNL SIKGTIDQID KTNNTYRQTI YVNPSGDNVI APVLTGNLKP NTDSNALIDQ QNTSIKVYKV DNAADLSESY FVNPENFEDV TNSVNITFPN PNQYKVEFNT PDDQITTPYI VVVNGHIDPN SKGDLALRST LYGYDSRFVW RSMSWDNEVA FNNGSGSGDG IDKPVVPEQP DEPGEIEPIP E | |
Greater than 90% as determined by SDS-PAGE. |