missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Foldase protein PrsA Protein

A DNA sequence encoding the Staphylococcus aureus (strain COL) Foldase protein PrsA, was expressed in the hosts and tags indicated.

€ 309.00 - € 3405.00

Specifications

Accession Number WP_000782121.1
Name S. aureus Foldase protein PrsA Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol prsA
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16090091
View Documents Promotion Details
enQuireBio™
QP7670-EC-100UG
100 μg
€ 985.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16010101
View Documents Promotion Details
enQuireBio™
QP7670-EC-1MG
1 mg
€ 3405.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16000101
View Documents Promotion Details
enQuireBio™
QP7670-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16020101
View Documents Promotion Details
enQuireBio™
QP7670-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16030101
View Documents Promotion Details
enQuireBio™
QP7670-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16040101
View Documents Promotion Details
enQuireBio™
QP7670-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

WP_000782121.1
Research Use Only
prsA
Recombinant Protein
E. coli
CGASATDSKE NTLISSKAGD VTVADTMKKI GKDQIANASF TEMLNKILAD KYKNKVNDKK IDEQIEKMQK QYGGKDKFEK ALQQQGLTAD KYKENLRTAA YHKELLSDKI KISDSEIKED SKKASHILIK VKSKKSDKEG LDDKEAKQKA EEIQKEVSKD PSKFGEIAKK ESMDTGSAKK DGELGYVLKG QTDKDFEKAL FKLKDGEVSE VVKSSFGYHI IKADKPTDFN SEKQSLKEKL VDQKVQKNPK LLTDAYKDLL KEYDVDFKDR DIKSVVEDKI LNPEKLKQGG AQGGQSGMSQ
Greater than 90% as determined by SDS-PAGE.
S. aureus Foldase protein PrsA Protein
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Product Certifications