missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Serine-aspartate repeat-containing protein 1
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Serine-aspartate repeat-containing protein E, was expressed in the hosts and tags indicated.
€ 309.00 - € 3405.00
Specifications
Accession Number | WP_000610237.1 |
---|---|
Name | S. aureus Serine-aspartate repeat-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | sdrE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011691
|
enQuireBio™
QP7933-EC-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031691
|
enQuireBio™
QP7933-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021691
|
enQuireBio™
QP7933-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041691
|
enQuireBio™
QP7933-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051691
|
enQuireBio™
QP7933-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061691
|
enQuireBio™
QP7933-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000610237.1 | |
Research Use Only | |
sdrE | |
Recombinant Protein | |
E. coli | |
AENTSTENAK QDDATTSDNK EVVSETENNS TTENDSTNPI KKETNTDSQP EAKEESTTSS TQQQQNNVTA TTETKPQNIE KENVKPSTDK TATEDTSVIL EEKKAPNYTN NDVTTKPSTS EIQTKPTTPQ ESTNIENSQP QPTPSKVDNQ VTDATNPKEP VNVSKEELKN NPEKLKELVR NDNNTDRSTK PVATAPTSVA PKRLNAKMRF AVAQPAAVAS NNVNDLITVT KQTIKVGDGK DNVAAAHDGK DIEYDTEFTI DNKVKKGDTM TINYDKNVIP SDLTDKNDPI DITDPSGEVI AKGTFDKATK QITYTFTDYV DKYEDIKARL TLYSYIDKQA VPNETSLNLT FATAGKETSQ NVSVDYQDPM VHGDSNIQSI FTKLDENKQT IEQQIYVNPL KKTATNTKVD IAGSQVDDYG NIKLGNGSTI IDQNTEIKVY KVNPNQQLPQ SNRIYDFSQY EDVTSQFDNK KSFSNNVATL DFGDINSAYI IKVVSKYTPT SDGELDIAQG TSMRTTDKYG YYNYAGYSNF IVTSNDTGGG DGTVKPEEK | |
Greater than 90% as determined by SDS-PAGE. |
S. aureus Serine-aspartate repeat-containing protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |