missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Staphopain B Protein
A DNA sequence encoding the Staphylococcus aureus Staphopain B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9880-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_001088793.1 | |
1 mg | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
S. aureus Staphopain B Protein | |
Research Use Only | |
sspB | |
Recombinant Protein | |
Yeast | |
DQVQYENTLK NFKIREQQFD NSWCAGFSMA ALLNATKNTD TYNAHDIMRT LYPEVSEQDL PNCSTFPNQM IEYGKSQGRD IHYQEGVPSY EQVDQLTKDN VGIMILAQSV SQNPNDPHLG HALAVVGNAK INDQEKLIYW NPWDTELSIQ DADSSLLHLS FNRDYNWYGS MIGY | |
Greater than 90% as determined by SDS-PAGE. |