missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salamander Leptin Salamander Protein
Click to view available options
Quantity:
1 mg
100 μg
200 μg
Unit Size:
100µg
1mg
200µg
Specifications
Specifications
| Name | Salamander Leptin Salamander Protein |
| Quantity | 200 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | By inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its activity in that test is similar to that of mouse leptin (see Salomon et al (2006) Protein Expression and Purifcation 47, 128–136). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Salamander |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | The sequence of the ten N-terminal amino acids was determined and was found to be Ala-Ile-Met-Val-Asp-Gln-Leu-Arg-Met-Asp |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction