missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 BA.2.75 S (GCN4-IZ) His Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN ).
Specifications
Specifications
| Accession Number | YP_009724390.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | M.W.-observed: 143-170 kDa, under reducing conditions., M.W.-theroretical: 138 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike RBD protein Human SARS-CoV-2 BA0.2.75 Spike RBD (Val16-Lys1211) (Thr19Ile, Leu24del, Pro25del, Pro26del, Ala27Ser, Gly142Asp, Lys147Glu, Trp152Arg, Phe157Leu, Ile210Val, Val213Gly, Gly257Ser, Gly339His, Ser371Phe, Ser373Pro, Ser375Phe, Thr376Ala, Asp405Asn, Arg408Ser, Lys417Asn, Asn440Lys, Gly446Ser, Asn460Lys, Ser477Asn, Thr478Lys, Glu484Ala, Gln498Arg, Asn501Tyr, Tyr505His, Asp614Gly, His655Tyr, Asn679Lys, Pro681His, Arg682Ser, Arg685Ser, Asn764Lys, Asp796Tyr, Gln954His, Asn969Lys, Lys986Pro, Val987Pro) (C-terminus) Accession # YP_009724390.1 GCN4-IZ 6-His tag (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction