missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 nsp2 His (N-Term) Protein

Product Code. 18660288 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18660288

Brand: Novus Biologicals™ NBP307083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE
Formulation PBS, pH 7.5
Gene ID (Entrez) 43740578
Molecular Weight (g/mol) MolecularWeight-theroretical: 72.81 kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Gene Alias Non-structural protein 2, NSP2, p65 homolog
Conjugate Unconjugated
Species SARS-CoV-2
Recombinant Recombinant
Form Purified
Research Category Infections (Virus Bacteria and Parasites)
Purity or Quality Grade >90%, by SDS-PAGE quantitative densitometry by Coomassie Blue Staining
Protein SARS-CoV-2 nsp2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.