missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike RBD Fc Avi-tag Protein
Biotinylated
Brand: R&D Systems™ AVI10499-050
This item is not returnable.
View return policy
Description
The biotin to protein ratio is greater than 0.7 as determined by the HABA assay. Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN ).Specifications
YP_009724390.1 | |
Bioactivity | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
Biotin | |
50 μg | |
SARS-CoV-2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction