missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike S1 Subunit Fc Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 His-tag (Catalog # 933-ZN ).
Specifications
Specifications
| Accession Number | YP_009724390.1 |
| Conjugate | Unconjugated |
| For Use With (Application) | Bioactivity |
| Quantity | 100 μg |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Synonym | SARS-CoV-2 |
| Target | SARS-CoV-2 |
| Buffer | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction