missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IL10 / Interleukin-10 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.
€ 352.00 - € 3175.00
Specifications
Accession Number | NP_001009327.1 |
---|---|
Gene ID (Entrez) | 443342 |
Name | IL10 / Interleukin-10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028701
|
enQuireBio™
QP9777-YE-100UG |
100 μg |
€ 1100.00
100µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068701
|
enQuireBio™
QP9777-YE-500UG |
500 μg |
€ 2650.00
500µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038701
|
enQuireBio™
QP9777-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058701
|
enQuireBio™
QP9777-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078701
|
enQuireBio™
QP9777-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048701
|
enQuireBio™
QP9777-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Especificaciones
NP_001009327.1 | |
IL10 / Interleukin-10 Protein | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |
443342 | |
Research Use Only | |
IL10 | |
Recombinant Protein | |
Yeast | |
SRDASTLSDS SCTHFPASLP HMLRDVRAAF GKVKTFFQMK DQLNSMLLTQ SLLDDFKGYL GCQALSEMIQ FYLEEVMPQA ENHGPDIKEH VNSLGEKLKT LRLRLRRCHR FLPCENKSKA VEQVKRVFNM LQERGVYKAM SEFDIFINYI ESYMTTKM | |
Greater than 90% as determined by SDS-PAGE. |