missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep IL10 / Interleukin-10 Protein

A DNA sequence encoding the Ovis aries (Sheep) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9777-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028701

  • € 860.00 / 100µg


Estimated Shipment
03-11-2020
Add to basket

Specifications

Specifications

NP_001009327.1
IL10 / Interleukin-10 Protein
100 μg
Not determined.
Full Length
Sheep
His
Tris-based buffer, 50% glycerol
443342
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL10
Recombinant Protein
Yeast
SRDASTLSDS SCTHFPASLP HMLRDVRAAF GKVKTFFQMK DQLNSMLLTQ SLLDDFKGYL GCQALSEMIQ FYLEEVMPQA ENHGPDIKEH VNSLGEKLKT LRLRLRRCHR FLPCENKSKA VEQVKRVFNM LQERGVYKAM SEFDIFINYI ESYMTTKM
Videos
SDS
Documents