missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep IL4 / Interleukin-4 Protein

Product Code. 16091710
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16091710 100 μg 100µg
16001720 10 μg 10µg
16041720 50 μg 50µg
16021720 200 μg 200µg
16031720 500 μg 500µg
16011720 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16091710 Supplier enQuireBio™ Supplier No. QP6224ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Ovis aries (Sheep) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001009313.2
Gene ID (Entrez) 443327
Name IL4 / Interleukin-4 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol IL4
Protein Length Full Length of Mature. Protein
Product Type Recombinant Protein
Cross Reactivity Sheep
Species E. coli
Protein Tag His-B2M
Sequence HKCDITLEEI IKTLNILTSR KNSCMELPVA DVFAAPKNAT EKETFCRAGI ELRRIYRSHM CLNKFLGGLD RNLSSLASKT CSVNEAKTST STLRDLLERL KTIMREKYSK C
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.