missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Toxoplasma Gondii Toxoplasma P32 Protein

A cDNA sequence encoding the Toxoplasma P32 was constructed and used to recombinantly synthesize the protein.

€ 238.00 - € 1815.00

Specifications

Name Toxoplasma Gondii Toxoplasma P32 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Toxoplasma Gondii
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15924709
View Documents Promotion Details
enQuireBio™
QP13788-100UG
100 μg
€ 238.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15944709
View Documents Promotion Details
enQuireBio™
QP13788-500UG
500 μg
€ 910.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15934709
View Documents Promotion Details
enQuireBio™
QP13788-1MG
1 mg
€ 1815.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Toxoplasma Gondii Toxoplasma P32 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Toxoplasma Gondii
His
Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Sterile Filtered solution containing containing PBS and 25mM arginine.
Videos
SDS
Documents

Documents

Product Certifications