missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Viral HPV E6 Protein
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
The Recombinant Viral HPV E6 Protein is derived from E. coli. The Recombinant Viral HPV E6 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P03126.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT. |
| Gene ID (Entrez) | 1489078 |
| Name | HPV E6 |
| Quantity | 25 μg |
| Source | E. coli-derived viral E6 protein Met1 - Leu158 with a N-terminal Ser-His Contains Cys to Ser substitutions at the following positions: 23, 58, 87, 104, 118, 147 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >98%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction