missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Parvovirus B19 VLP VP1 Protein
Click to view available options
Quantity:
1 mg
100 μg
5 mg
Unit Size:
100µg
1mg
5mg
Specifications
Specifications
| Name | Virus Parvovirus B19 VLP VP1 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Virus |
| Species | E. coli |
| Protein Tag | Untagged |
| Buffer | Parvovirus B19 VLP VP1 protein solution is supplied in PBS pH 7.4 and 0.02% Sodium azide. |
| Purity or Quality Grade | Greater than 90.0% as determined by 12% PAGE (coomassie staining). |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction