missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Perth 16/09 Protein
A cDNA sequence encoding the Perth 16/09 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 1665.00
Specifications
Name | Virus Perth 16/09 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15922239
|
enQuireBio™
QP13010-1UG |
1 μg |
€ 137.00
1µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942239
|
enQuireBio™
QP13010-5UG |
5 μg |
€ 212.00
5µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932239
|
enQuireBio™
QP13010-50UG |
50 μg |
€ 1665.00
50µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Virus Perth 16/09 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
His | |
The H3N2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPMQKLPGN DNSTATLCLG HHAVPNGTIV KTITNDQIEV TNATELVQSS STGEICDSPH QILDGKNCTL IDALLGDPQC DGFQNKKWDL FVERSKAYSN CYPYDVPDYA SLRSLVASSG TLEFNNESFN WTGVTQNGTS SACIRRSKNS FFSRLNWLTH LNFKYPALNV TMPNNEQFDK LYIWGVHHPG TDKDQIFLYA QASGRITVST KRSQQTVSPN IGSRPRVRNI PSRISIYWTI VKPGDILLIN STGNLIAPRG YFKIRSGKSS IMRSDAPIGK CNSECITPNG SIPNDKPFQN VNRITYGACP RYVKQNTLKL ATGMRNVPEK QTRHHHHHH | |
Greater than 90% as determined by SDS-PAGE. |