missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Virus Perth 16/09 Protein

Product Code. 15942239
Change view
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15942239 5 μg 5µg
15922239 1 μg 1µg
15932239 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15942239 Supplier enQuireBio™ Supplier No. QP130105ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Perth 16/09 was constructed and used to recombinantly synthesize the protein.

Specifications

Name Virus Perth 16/09 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Virus
Species Baculovirus
Protein Tag His
Sequence ADPMQKLPGN DNSTATLCLG HHAVPNGTIV KTITNDQIEV TNATELVQSS STGEICDSPH QILDGKNCTL IDALLGDPQC DGFQNKKWDL FVERSKAYSN CYPYDVPDYA SLRSLVASSG TLEFNNESFN WTGVTQNGTS SACIRRSKNS FFSRLNWLTH LNFKYPALNV TMPNNEQFDK LYIWGVHHPG TDKDQIFLYA QASGRITVST KRSQQTVSPN IGSRPRVRNI PSRISIYWTI VKPGDILLIN STGNLIAPRG YFKIRSGKSS IMRSDAPIGK CNSECITPNG SIPNDKPFQN VNRITYGACP RYVKQNTLKL ATGMRNVPEK QTRHHHHHH
Buffer The H3N2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.