missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Yeast GLRX1 Yeast Protein

Product Code. 15989018
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15989018

Brand: enQuireBio™ QP1199010ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GLRX1 Yeast was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 850322
Name GLRX1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol GRX1
Product Type Recombinant Protein
Cross Reactivity Yeast
Species E. coli
Protein Tag His
Sequence MVSQETIKHV KDLIAENEIF VASKTYCPYC HAALNTLFEK LKVPRSKVLV LQLNDMKEGA DIQAALYEIN GQRTVPNIYI NGKHIGGNDD LQELRETGEL EELLEPILAN
Buffer Glutaredoxin solution contains 25mM Tris-HCl pH 7.5 and 0.01% Na Azide.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.